Molex Ratchet Crimp Tool, Newark/Element
Supplier: NEWARK/ELEMENT 14 TE
63819-1000
76253-836EA 1484.43 USD
76253-836
Molex Ratchet Crimp Tool, Newark/Element
Crimpers, Cappers and Decappers
This full cycle ratcheting hand tool ensures complete crimps and has ergonomic soft grip handles for comfortable crimping.
  • Right and left-handed applications
  • For Mini-Fit™ Jr. male and female crimp terminals, 22 to 28AWG
A precision user-friendly terminal locator wire stop holds terminals in the proper crimping position.
Certifications: Tool is IPC/WHMA A-620 Class 2 compliant.