HY-PRO® NRT® Taps, Sid Tool Co
Supplier: SID TOOL CO INC TE
9247875
76253-800EA 88.65 USD
76253-800
HY-PRO® NRT® Taps, Sid Tool Co
Tool Sets
Forming tap with UNC thread.
  • Tin coated
  • Thread limit H2
  • Threads 40 per inch
  • Chamfer 2.5 P