Retention Tester, DMC
Supplier: DMC TE
HT250-1
76253-648EA 326.79 USD
76253-648
Retention Tester, DMC
Tool Sets
Ths testers to test retention of socket contacts, use the socket tester tip with a preset tool insert tester (pin) into mating end of contact.
Tool must be in a straight line with contact. For pin contacts, fit pin tester to tool body and place over contact at the mallng end.