Antisplash Membranes, JBC Tools USA
Supplier: JBC TOOLS USA INC. TE
CL7882
76253-642EA 46.52 USD
76253-642
Antisplash Membranes, JBC Tools USA
Supports Support Sleeves
Antisplash rubber to prevent splashing of solder particles and maintain the work area clean.
  • Weight: 8 g (0.28 oz.)