787MS-SS MicroSpray™ Valves, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
7029409
76253-640EA 8006.96 USD
76253-640
787MS-SS MicroSpray™ Valves, EFD
Valves Pressure-Relief Valves
The 787MS-SS MicroSpray™ valve’s innovative design incorporates a small gauge disposable dispensing tip to apply a precise, uniform spray pattern more than 30% smaller than standard spray valves.
  • High transfer efficiency
  • No overspray or mist
  • Consistent spray pattern
  • Faster throughput
  • Cycle rate exceeds 400 cycles per minute
  • Accommodates tip centering guide
Using Low Volume Low Pressure (LVLP) technology it produces spray patterns as small as 3.3 mm (0.13") and as great as 19 mm (0.75") in diameter. A small gauge, 0.3 mm (0.013") to 0.1 mm (0.004"), dispense tip replaces the standard spray nozzle, concentrating the LVLP air used to atomize coatings into smaller spray patterns.
The 787MS-SS increases throughput and allows greater process control, while reducing production costs and rejects. The valve needle seats in the hub of the dispensing tip, virtually eliminating dead fluid volume between shots.
Integration with an EFD automated dispensing system for even faster throughput.
Delivery information: Kit includes spray tip kit, air hoses, fluid inlet fitting, barrel reservoirs, and adapter assembly for reservoir pressure.