Hand Soldering Tweezer Cartridge, Blade, OK International
Supplier: OK INTERNATIONAL TE
TCP-BLH50 TCP-BLP1 TFP-BLP1
76245-642EA 374.85 USD
76245-642 76245-658 76253-542
Hand Soldering Tweezer Cartridge, Blade, OK International
Soldering Supplies Soldering Tips
Precision tweezer cartridge used with the MFR single and dual platforms (MFR-1100 and MFR-2200) with the MFR-H4-TW tweezer hand-piece. Removes even the smallest of discrete components, such as 0201 packages. Second digit denotes substrate material (damage tolerance).