Proto® Socket Bit, Grainger
Supplier: GRAINGER
J47705/64
76253-508EA 44.26 USD
76253-508
Proto® Socket Bit, Grainger
Tool Sets
The Proto® socket bit has a hex opening that allows the bit holder to take the torque load.
  • Polished chrome finish
  • Insert length: 1"
Made from a heat-treated alloy for wear resistance which minimizes distortion for a long life.