Dispensing Gun, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
7023125
76253-466EA 536.88 USD
76253-466
Dispensing Gun, EFD
Fluid Adhesive Dispensers and Pistons
Ideal for touch-ups, low-volume assembly and field work. They can be used with all EFD syringe barrels, pistons and tips.
  • Ergonomic design
  • Fatigue-free dispensing of thick fluids
  • Positive shutoff, no dripping
  • Simplified maintenance
  • Reusable
Features 10:1 mechanical leverage that makes it easy to dispense thick materials like greases and silicones without hand fatigue. A clean cutoff when the trigger is released prevents oozing between fluid applications.