Metcal™ Precision Tweezer Workstand, Ok International
Supplier: OK INTERNATIONAL TE
MFR-WSPT
76253-322EA 277.74 USD
76253-322
Metcal™ Precision Tweezer Workstand, Ok International
Soldering Supplies Soldering Stations
For use with Metcal™ precision tweezer hand-piece on MFR series systems.
  • Comes with rectangular sponge and brass pad to keep your solder tips clean
  • Auto-sleep function to save tip life when not in use