Pneumatic Applicator with Dual Cartridge, Zoro
Supplier: ZORO - CUSTOMER SERVICE TE
G6123625
76253-286EA 1423.33 USD
76253-286
Pneumatic Applicator with Dual Cartridge, Zoro
Fluid Adhesive Dispensers and Pistons
This is for pneumatic adhesive dispensing, 1:1 and 2:1 mix ratio products in 50 ml Dual cartridge.