Crimp Tool with Y501 Die Set, DMC
Supplier: DMC TE
HX4-501
76253-222EA 2961.13 USD
76253-222
Crimp Tool with Y501 Die Set, DMC
Crimpers, Cappers and Decappers
This open frame crimp tool is comes with pre-installed Y501 die set.
  • For use with pre-insulated terminal crimp 26-10 AWG