Wire Marking Sleeves, PermaSleeve®, Brady
Supplier: Brady Worldwide
142096
76253-218EA 713.73 USD
76253-218
Wire Marking Sleeves, PermaSleeve®, Brady
Labels
These PermaSleeve® PS heat-shrink polyolefin (B-342) wire marking sleeves combine speed, ease of use, and permanence to create long-lasting, high quality identification.
  • Fade-resistant and smudge-proof for superior aesthetic appearance
  • With thermal transfer print technology
  • Cold resistant and flame-retardant with an operating temperature of –55 to 135 °C (–67 to 275 °F)
These heat shrinkable sleeves fit snugly around wires for maximum insulation protection and permanent identification.
Certifications: Meets SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202; RoHS Compliant; and UL Recognized.