QuadraTest Multimode Electronic Test Indicator, Sid Tool
Supplier: SID TOOL CO INC TE
20426136
76253-182EA 3883.73 USD
76253-182
QuadraTest Multimode Electronic Test Indicator, Sid Tool
Measuring and Leveling Tools
Multimode electronic test indicator offers unrivaled measuring versatility in an electronic test indicator!
Available with a 1/2" or 17/16" contact point. The QuadraTest is the perfect solution to test indicating applications.
Features four measuring modes (Minmum, Maximum, TIR and direct), with an RS-232 interface that is water, oil and coolant resistant to IP65 compliancy with or without the interface cable installed! The QuadraTest also features true inch/metric conversion, a 11/2" diameter dial face that rotates a full 360° with both analog bar graph and numeric digital display.
Certifications: Meets or exceeds DIN/ISO standards.