Taper Shank, CAT40, Sid Tool Co
Supplier: SID TOOL CO INC TE
80263767
76253-136EA 540.55 USD
76253-136
Taper Shank, CAT40, Sid Tool Co
Tool Sets
CAT40 Taper Shank, with end mill holder.
  • DIN flange coolant through
  • Through-spindle coolant
  • Maximum TIR 0.0002″ OD to ID
Shank traverse ground to obtain optimum accuracy.
Certifications: Meet or exceed ANSI specifications.
Delivery information: Each tool comes with inspection lab certificate.