Statfree B2® Static Dissipative Two Layer Floor Mats, Menda
Supplier: Desco
66177
76253-010EA 174.71 USD
76253-010 76253-808
Statfree B2® Static Dissipative Two Layer Floor Mats, Menda
Floor Mats Permanent Floor Mats
Statfree B2® heat fused thermoplastic two layer vinyl provides reliable path to ground and eliminates static charge generation as a worksurface table mat, shelf liner and cart liner.
  • Lo-Char flame retardant additive
  • Laser engraved with ESD protective symbol and resistance
Dissipative (1×106 to <1×109 ohms Rtt) top color layer and conductive (<106 Rtt) bottom black layer.
Certifications: Meets ANSI/ESD S20.20, ANSI/ESD S4.1.