Turrets for Crimp Tools, DMC
Supplier: DMC TE
TP465
76252-772EA 345.34 USD
76252-772
Turrets for Crimp Tools, DMC
Crimpers, Cappers and Decappers
Single position head is used with the M22520/1-01 (AF8) tool frame and adapts the tool frame to a particular application.
The data plate on each positioner designates which contacts the positioner accommodates for its wire size and indicates selector position.
Certifications: Qualified to AS22520/1.