Complete SMD Hot Air Rework System, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FR811-SET
76252-738EA 18884.81 USD
76252-738
Complete SMD Hot Air Rework System, American Hakko Products
Soldering Supplies Soldering Stations
A complete, economical SMD hot air rework system, including the HAKKO FR-811 hot air station, bottom heater, handpiece rework fixture, board holder and vision system.
  • Manual or Automatic Operating Modes
  • 6-User Programmable Presets
  • Turbine Airflow
  • Features provide stabilization for accuracy when reworking small or difficult components
This set can be transformed into a complete SMD rework system with added accessories such as bottom heater, board holder and handpiece fixture.