NonWoven Sponge, McKesson
Supplier: MCKESSON MEDICAL-SURGICAL INC TE
16-40334
76252-608PK 11.51 USD
76252-608
NonWoven Sponge, McKesson
Wipes Sponges
McKesson non-woven sponges with high absorbency.
  • Non sterile
  • Poly/rayon blend
  • High-quality alternative to cotton gauze
  • Strong and durable
  • Excellent absorption with virtually no linting
  • Greater wicking properties than cotton gauze
  • Single use
Appropriate for wound dressing, cleaning, prepping, and scrubbing.
Packaging: 200 per pack, 20 packs per case.