FiberChek™ Probe Microscope, Fiber Instrument Sales
Supplier: FIBER INSTRUMENT SALES INC. TE
FIT-FC-KIT2
76252-258EA 16900.57 USD
76252-258
FiberChek™ Probe Microscope, Fiber Instrument Sales
Microscopes
FiberChek probe equips fiber technicians with a single autonomous, handheld solution they can rely on for all of today’s fiber inspection needs that delivers fast and accurate performance.
  • Integrated touch screen with live fiber viewing
  • Auto-center and auto-focus
  • Built-in fiber end-face analysis
  • User-selectable acceptance profiles
  • Results storage on device
Integration with iOS, Android™, Windows devices via WiFi, Bluetooth™, and USB.