SMD Hot Air Rework System, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FR811-04
76252-244EA 15334.29 USD
76252-244
SMD Hot Air Rework System, American Hakko Products
Soldering Supplies Soldering Stations
An advanced SMD rework station with vacuum pickup and a multitude of features for every need. When integrated with a bottom heater, board fixture and vision system, the FR-811 is a complete SMD rework system and the ideal choice for bottom terminated components.
  • New graphic interface display and navigation system that simplifies operation
  • Dual T/C inputs allow users to monitor part and board temperatures while controlling the ramp rate for a safe, accurate and repeatable process
  • Automatic mode gives users the ability to fully program profiles with up to 6 stages
  • Onboard storage of up to 50 profiles with an external USB port for flash drive memory for storage of profiles
  • A dedicated USB port can also be used to integrate the FR-811 with a PC using the included software
  • Integrated vacuum pickup with a visual indicator in the handpiece that indicates when the component has been completely reflowed
Features dual thermocouple inputs for part and board monitoring, PC integration via USB port with software, an additional external USB port for flash drive memory for storage of profiles, onboard storage of up to 50 profiles, manual and auto modes, auto teach mode for profiling, password lockout, and lots more.
Ordering information: Nozzles (N51 Series) has to be ordered.