Eyepiece, LX Microscopes by Unitron
Supplier: LX Microscopes by UNITRON
23762
76252-216EA 411.25 USD
76252-216
Eyepiece, LX Microscopes by Unitron
Microscope Eyepieces
This is for use with LX Microscopes by UNITRON systems 273, 373 and VIP.
  • Super wide 23 mm field of view
  • Shipping weight 1 lbs.