Filter Bags, National Filter Media Corp
Supplier: NATIONAL FILTER MEDIA CORPT TE
0509016-196-21
76252-152EA 348.03 USD
76252-152
Filter Bags, National Filter Media Corp
Bags Blender Bags
Dust collector filter bags with PE felt media.
  • One circular sewn end
  • One open end