FEP Tubing, Newage Industries
Supplier: NewAge Industries
FEP250-030-100
76251-790EA 629.28 USD
76251-790
FEP Tubing, Newage Industries
Tubing Transfer Tubing
High purity solution for ultra-clean applications and are excellent with corrosive media.
  • Flexible but will kink if bent too tightly
  • Can be welded and sealed by melting
  • Very tough material, can be rigorously cleaned
  • For visual clarity FEP is an alternate to PTFE
They resist combustion and do not promote flame spread. They are also approved for use in food contact applications.
Tolerance of ±0.005" for both int.Ø and ext.Ø, wall tolerance of ±0.007"
Certifications: UL 94 VO flammability rated, compliant with FDA: 21 CFR 177.1550.