Precision Potentiometer, Digi-Key
Supplier: DIGI-KEY CORPORATION
3590S-2-103L
76251-628EA 77.38 USD
76251-628
Precision Potentiometer, Digi-Key
Electrical Meters Voltmeters
Precision Potentiometer with plastic or metal shaft and bushings.
  • Wirewound
  • Solder lugs or PC pins
  • Sealable (Full body seal)
Designed for use in HMI applications and with optional AR pin feature.
Certifications: RoHS compliant.