Disposable Pinch Tube Pen, TS1201, OK International
Supplier: OK INTERNATIONAL TE
1201-000-004PK
76251-616EA 150.96 USD
76251-616
Disposable Pinch Tube Pen, TS1201, OK International
Soldering Supplies Soldering Tips
he TS1201 is a hand-held disposable pinch tube pen that provides control for continuous or micro shot applications of low to semi-viscous fluids. Once the dispense cycle is complete, the operator releases the trigger handle and the internal spring forces the shut-off piston to "pinch" the disposable feed path,
preventing further fluid flow. The TS1201 is fitted with a built in flow rate adjustment.
  • Disposable "pinch" tube design
  • Simple to operate
  • Compact size and weight
The only part of the valve making contact with the fluid being dispensed is the disposable pinch tube assembly making the TS1201 ideally suited to dispensing 2-part and CA (Cyanoacrylate) fluids. Black pinch tubes are made available for use with UV adhesives.
The disposable pinch tubes come pre-assembled with a male Luer lock connection for quick and easy tip attachment.
Delivery information: Includes 4 pinch tubes (2 clear and 2 black), 6 luer lock fluid lines (3 clear and 3 black), hex keys, dispensing tip selection pack.