Splicing Tool, Splicetronics Solutions
Supplier: SPLICETRONICS SOLUTIONS INC TE
ST410
76251-566EA 1139.25 USD
76251-566
Splicing Tool, Splicetronics Solutions
Tool Sets
Stapler style splice tool designed specifically for SMT carrier tapes. Used with the splice clip/shim frame and splice tapes of any width. Ensures SMT component reels can be joined quickly and securely.
  • Slider to push splice clips forward
  • Carbon steel
Delivery information: It includes clamps to secure reels.