Amp-TE Connectivity Ratchet Crimp Tool, Newark/Element
Supplier: NEWARK/ELEMENT 14 TE
59250
76251-548EA 5558.18 USD
76251-548
Amp-TE Connectivity Ratchet Crimp Tool, Newark/Element
Crimpers, Cappers and Decappers
This T-head hand crimp tool is designed for AMP PIDG/STRATO-THERM/PLASTI 26-14AWG terminals and splices.
  • The handle of the crimp tool is color-coded to match the color coding of the product to be applied
  • PLASTI-GRIP terminals on solid or stranded copper wire sizes 22 through 14AWG
  • STRATO-THERM terminals on stranded copper wire sizes 26 through 14AWG
The insulation adjustment indicator is used to control crimp height of the insulation barrel. It can be set at any of four positions corresponding to insulation diameter.
The CERTI-CRIMP hand crimping tool ratchet control ensures full crimping of the product. Once engaged, the ratchet will not release until the handles have been fully closed.