Gauges, Jensen Tools
Supplier: TESTEQUITY LLC TE
ATG36Z-S
76251-394EA 1863.32 USD
76251-394
Gauges, Jensen Tools
Wrenches
Analog handheld torque gauge ide for measuring and tightening in low torque ranges.
  • Three Fingered Keyless Metal Chuck
  • CW and CCW measuring with side and top memory pointers
Delivery information: Calibration Certificate Included.