PharmaMOP® Cleaning Kits, Foamtec
Supplier: FOAMTEC INTERNATIONAL, LLC.
FS4500SAHARA
76251-300KT 363.27 USD
76251-300
PharmaMOP® Cleaning Kits, Foamtec
Mops
PharmaMOP® is designed for cleanroom cleaning SOP. The light weight, 360 degree rotating and locking mop head give operators excellent control cleaning floors, walls and ceilings. The ultra-light handle, wringer less bucket system and drip resistant mop heads allow operators to efficiently carry out SOP’s with improved ergonomics.
PharmaMOP® can be used withgure eight, front to back or top to bottom motions and the rotating mop frame enable cleaning of tight spaces. The PharmaMOP® handle and SingleSTEP™ mop heads have been designed to allow for greatly improved cleaning efficiency of soils and residues from difficult to clean textured floors while at the same time allowing ease of mopping. The entire PharmaMOP® system is compatible with steam sterilization and all of the major disinfectants.
Delivery information: It includes one mop and 10 mop heads.