Electric Screwdrivers, HIOS CL, Mountz
Supplier: MOUNTZ INC TE
144116 144121
76251-266EA 3471.13 USD
76251-266 76251-268
Electric Screwdrivers, HIOS CL, Mountz
Screwdrivers
The CL-Series electric torque screwdriver is designed for heavy-duty production and delivers reliability as well as precision torque.
  • Low DC voltage for safe operation and long life
  • Precision automatic shut-off clutch once preset torque is achieved
  • Compact size with a grip designed for operator comfort
  • External torque adjustment scale
  • External brush replacement means no downtime for brush replacement
  • Torque levels can be easily adjusted by turning down the external adjustment ring
  • Connectable with the Scout screw counter
Accurate torque control provides precise and controlled fastening with automatic shut-off clutch. The full auto shut-off clutch on this electric screwdriver completely stops the bit from turning once the torque setting is reached. This feature protects the fastener from being damaged by the electric screwdriver's bit.
Common usages for this electric screwdriver include demanding assembly-fastening applications that require torque control, screw counting functionality and ESD protection.
Certifications: Drivers meet electrical safety standards in the U.S. and Europe (UL Listed and CE Certified).