Shop Air Desoldering and Soldering Station, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FM205-01
76251-120EA 1886.29 USD
76251-120
Shop Air Desoldering and Soldering Station, American Hakko Products
Soldering Supplies Soldering Stations
FM-205 is a “shop-air” desoldering and soldering station. The shop-air feature provides for higher vacuum pressure and increased suction flow in comparison to the FM-204.
  • Digital Temperature Control
  • Uses composite nozzles for improved thermal performance
  • Quick-change composite nozzles (N3 series)
  • Uses low cost disposable filter cartridges
  • Ejector vacuum generator
Can be used as a soldering station with a Hakko FM2027 connector. This is less prone to clogging as compared to older desoldering stations when used on lead free applications.
Delivery information: Supplied with low temperature alarm, sleep mode functionality and auto power off.