Phillips® Power Bits, Wiha Tools
Supplier: WIHA TOOLS TE
74106
76251-078PK 186.16 USD
76251-078 76251-732
Phillips® Power Bits, Wiha Tools
Screwdrivers
The ideal multi-purpose bits for all around quality and performance.
  • Highly resistant to wear and tear for a long tool life
  • High grade CVM steel, hardened torsion zone to prevent uneven torque distribution
These are suitable for all applications for both manual and machine Hardness HRC 59-61.
Certifications: DIN 3126 ISO 1173.