Klein® Cable Cutter, TESTEQUITY LLC TE
Supplier: TESTEQUITY LLC TE
63050
76250-926EA 99.88 USD
76250-926
Klein® Cable Cutter, TESTEQUITY LLC TE
Cutters
High leverage design and shear-type jaws cut up to 4/0 aluminum, up to 2/0 soft copper, and up to 100 pair, 24 AWG communication cable.