Minigrabber® Test Clips and Kits, Digi-Key
Supplier: DIGI-KEY CORPORATION
5522
76250-862PK 120.5 USD
76250-862
Minigrabber® Test Clips and Kits, Digi-Key
Cables
Do-It-Yourself Minigrabber® test clip for maximum 0.090" wire diameter.
  • Contacts: Gold plated Beryllium Copper
  • Spring: Stainless steel
  • Insulation: Glass filled nylon
Delivery information: Kit includes one each of ten colors of Do It Yourself (DIY).