Smoke Absorber, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FA400-04
76250-806EA 267.17 USD
76250-806
Smoke Absorber, American Hakko Products
Fume Extractors
The FA-400 bench top smoke absorber is a quick, safe, and efficient way to absorb soldering smoke and fumes. Optional high efficient filters are available and all filters are easily replaceable.
  • Can be used in a vertical (standing up) position or low-profile position (laying down)
  • Unit is made of non-sloughing static dissipative conductive material and operates quietly
  • ESD-safe by design
Standard activated carbon filter absorbs up to 80 percent of the odors from the air and is easy to replace.