Adapters, Digi-Key
Supplier: DIGI-KEY CORPORATION
BPJJ02AUX
76250-608EA 25.46 USD
76250-608
Adapters, Digi-Key
Electrical Sockets
Adapter connector Phono (RCA) jack to Phono (RCA) jack panel mount.
  • Brass body with gold finish
  • Nickel plated contact material
  • Red insulation
  • Unshielded
Female to female connector.
Delivery information: Includes 1 connector, 1 nut and 1 washer.