Chicago Latrobe® NAS-Type Heavy-Duty Jobber Length Drills, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
41737
76250-282PK 154.9 USD
76250-282 76253-666
Chicago Latrobe® NAS-Type Heavy-Duty Jobber Length Drills, Cutting Tool Supply
Tool Sets
Heavy-duty construction for drilling in tougher materials and 135° P3 split point is self-centering for reduced thrust and easier penetration.
  • Made of premium high-speed steel
Black oxide surface finish increases wear resistance and adds lubricity to improve chip flow.
Certifications: Manufactured to NAS 907 Type B geometry aerospace specifications.