Power Relay, Digi-Key
Supplier: DIGI-KEY CORPORATION
G6CU-1114P-US-
76250-132EA 38.16 USD
76250-132
Power Relay, Digi-Key
Power Supplies
This general purpose coil through hole power relay is designed with cadmium free, silver alloy contact material.
  • Operate time: 10 ms
  • Coil resistance: 125 Ω
Coil voltage and current is measured as 5 V and 40 mA respectively.