Go/No Go Plug Gage Assembly, Zoro
Supplier: ZORO - CUSTOMER SERVICE TE
301110040
76250-088EA 372.9 USD
76250-088
Go/No Go Plug Gage Assembly, Zoro
Measuring and Leveling Tools
Go/No Go, Taperlock thread plug gage, Class 2B.
  • Made of tool steel
  • Chrome plated finish
  • Tolerance Class X
  • UNC Thread
For calibration of measuring equipment or to gage parts directly.
1.75" (handle)
Delivery information: Includes Certificate of Accuracy Traceable to NIST.