Heavy Duty Work Stand, CF Clark
Supplier: CF CLARK ASSO. INC. TE
WS248
76249-956EA 803.31 USD
76249-956
Heavy Duty Work Stand, CF Clark
Furniture Tables and Workbenches
Durable flush mounted 12 gauge steel shelves.
  • Clearance between shelves is 20"
  • Clearance under bottom shelf is 7"
Corner posts 1¹/₂ by ³/₁₆" thick angle with pre punched mounting pads.