Micro Soldering Iron Handpiece, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FM2032-51
76249-942EA 360.32 USD
76249-942
Micro Soldering Iron Handpiece, American Hakko Products
Soldering Supplies Soldering Irons
Add precision micro soldering capabilities for soldering and rework system. This slim, lightweight handpiece has a tip-to-grip distance optimized for operator dexterity while working under microscopes.
  • Thin, burn resistant iron cord
  • Quick-change T30 Series tips
  • ESD Safe
Cool running grip even when operating at 450 °C for extended periods.
Delivery information: Supplied with micro soldering handpiece.