Kevlar® Cutter with Blue Grips, Swanstrom Tools
Supplier: SWANSTROM TOOLS USA TE
9180
76249-846EA 63.42 USD
76249-846
Kevlar® Cutter with Blue Grips, Swanstrom Tools
Cutters
Designed and manufactured for cutting Kevlar® and other high strength fibers.
  • Abrasion resistance and stretch free nature
Handles are free of confining finger loops and return spring brings tool back to an open position after each cut. one of the blades is serrated and acts to prevent the fibers from sliding out of the front of the scissor when cutting.