Tip Detector, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
7360893
76249-810EA 7248.19 USD
76249-810
Tip Detector, EFD
Leak Detection Systems Liquid Leak Detection Systems
Tip detector allows to automatically update both the XY offsets and the Z height by clicking on needle Z detect in the EV series automated dispenser systems.
  • The needle Z detect button is present only on systems that include the tip detector