Turbo Mixer™ 295 Series, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
7701836
76249-808EA 7.54 USD
76249-808
Turbo Mixer™ 295 Series, EFD
Stirrers Stirring Tools
Turbo Mixer feature a patented design that consists of alternating left- and right-hand elements with intermittent flow inverters that channel
fluids from the walls into the center of the mixer and from the center to the walls for thorough, complete blending.