Unity™ HiTemp™ Syringe Barrels, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
7360473 7360475
76249-738PK 219.28 USD
76249-738 76249-740
Unity™ HiTemp™ Syringe Barrels, EFD
HiTemp™ barrels withstand 125 °C at up to 6 bar. These syringes can be used with all hot melt adhesives, including moisture sensitive polyurethanes (PURs) and silyl modified polymers (SMPs).
  • Proprietary materials that maintain integrity at high temperatures
  • 30 ml capacity compatible with most industry hot melt systems
  • Higher temperatures allow for narrower beads and lines for more precise dispensing
Extreme HiTemp™ barrels withstand 180 °C at up to 7 bar. These syringes can be used with polyamides
and other types of hot melt materials. They also block all UV light between 200 and 500 nm.