Shielding Bag, SCS Division of Desco, Desco Industries
Supplier: Desco
8171518
76249-724PK 262.4 USD
76249-724 76249-726
Shielding Bag, SCS Division of Desco, Desco Industries
Bags Sample Bags
Static Shield Bag 81705 Series are manufactured from four layers static dissipative coating, polyester, metal and polyethylene laminate.
  • The polyester dielectric in concert with the metal layer provides discharge shielding
The exterior is static dissipative and allows electrostatic charges to be removed when grounded.