NETcat® Micro NC-100 Digital Voice, Data and Video Wiring Tester, Jensen Tools
Supplier: TESTEQUITY LLC TE
NC-100
76249-716EA 477.1 USD
76249-716
NETcat® Micro NC-100 Digital Voice, Data and Video Wiring Tester, Jensen Tools
Cable Testers
The NETcat® Micro is a device that verifies the integrity of both twisted pair and coaxial telecom cables. It uses high-speed digital technology to verify wiring continuity and check for the correct allocation of wire pairs, showing the results on a backlit display.
  • Tests shielded twisted pair (STP), unshielded twisted pair (UTP) and coaxial cables
  • TIA/EIA standard wiring pair verification
  • Complete set of connector types standard (telephone, LAN and coax)
  • Nested remote allows end-to-end testing of all wiring types
  • Detection of common wiring and connector errors
  • Warning of hazardous voltages
  • Checks for reversed telephone wires
Four durable keys select the test and wire tracing modes. A wide range of tracing tone options makes it easy to track and identify high performance Category 5/6/7 wiring, and the tracing tones are compatible with Greenlee’s 200XP noise-immune tone probe. An auto-off function helps extend battery life.
Generates three distinct, selectable digital tone patterns for cable tracing and troubleshooting. Detects shorts, opens, reversed polarity, crossed and split pairs. Tracing modes can be adjusted for high performance LAN cables.