Pin Gage, Plus, Cylindrical, Zoro
Supplier: ZORO - CUSTOMER SERVICE TE
911105000
76249-656EA 11.85 USD
76249-656
Pin Gage, Plus, Cylindrical, Zoro
Measuring and Leveling Tools
Pin Gage, Plus, Cylindrical, Class ZZ.
  • Black oxide coated
  • Made of 52100 tool steel
The gage has a accuracy of +0.0000/–0.0002".
Delivery information: Includes Certificate of Accuracy with NIST traceability.