Water for irrigation (WIR), sterile

Supplier: MCKESSON MEDICAL-SURGICAL INC TE

Synonyms: Dihydrogen oxide

14045
76249-620CS 255.66 USD
76249-620
Water for irrigation (WIR), sterile
Water

Irrigation Solution - Sterile Water for Irrigation


  • NDC number: 00338-0004-04
  • Latex Free Indicator: Not Made with Natural Rubber Latex


Caution: Not for Injection

Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR