Rotary Collet Magnet Wire Stripper, 27F, Jensen Tools + Supply
Supplier: TESTEQUITY LLC TE
M27F-C
76249-560EA 2698.71 USD
76249-560
Rotary Collet Magnet Wire Stripper, 27F, Jensen Tools + Supply
Cutters
The model 27F is designed to strip magnet wire, coils and resistor leads to within 0.015" (depending on insert style) of the component body. The rotating slotted stripping insert removes the film insulation leaving a clean strip with a square shoulder.
  • Standard with a durable powder coat finish
Strip lengths with the stop rod installed are up to 0.625" or with the stop rod removed up to 2.87". Unique stripping inserts are easily changed for various wire gauges and are available in hardened steel or carbide. With the optional flexible shaft assembly the model 27F can be converted from a bench top machine to a more portable unit able to process leads on heavy coils.